Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (human genomic)

Supplier: Promega Corporation

G1471
PAG1471EA 258.83 USD InStock
PAG1471
DNA (Deoxyribonucleic acid) (human genomic)
DNA (Deoxyribonucleic acid)
Human Male Genomic DNA is purified and stored in 10mM Tris-HCl (pH 8.0), 1mM EDTA >90% of the DNA is longer than 50kb in size as measured by pulsed-field gel electrophoresis. The DNA is suitable for Southern blot hybridizations, genomic analysis (including PCR) and genomic library construction. Human Genomic DNA comes from multiple anonymous donors.

>90% of the DNA is longer than 50kb in size as measured by pulsed-field gel electrophoresis. The DNA is suitable for Southern blot hybridizations, genomic analysis (including PCR) and genomic library construction. Human Genomic DNA comes from multiple anonymous donors.


Storage Temperature: Refrigerator
MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty