Would you like to log in again?
CellExp™ Human Recombinant Granzyme B (from HEK293 cells)
Supplier: BioVision
CellExp™
AB285779-10UG
10009-582EA 1195.09 USD InStock
10009-582
CellExp™ Human Recombinant Granzyme B (from HEK293 cells)
Proteins and Peptides
>95% Pure Active Human cell expressed Human Recombinant Granzyme B
Measured by its ability to cleave a peptide substrate, t-Butyloxycaronyl-Ala-Ala-Asp-ThioBenzyl ester (Boc-AAD-SBzl), in the presence of 5, 5’-Dithio-bis (2-nitrobenzoic acid) (DTNB). The specific activity is more than 800 pmol/min/µg.
Granzyme B (GZMB) also known as Cathepsin G-like 1 (CTSGL1), Cytotoxic T-lymphocyte proteinase 2 (Lymphocyte protease), Fragmentin-2, Granzyme-2, Human lymphocyte protein (HLP), SECT, CGL1, CSPB, CTLA1, GRB, which belongs to the peptidase S1 family. Granzyme subfamily GZMB / CTSGL1 contains 1 peptidase S1 domain. The catalytic activity of Granzyme B is “Preferential cleavage: -Asp-|-Xaa- >> -Asn-|-Xaa- > -Met-|-Xaa-, -Ser-|-Xaa-“, and is inactivated by the serine protease inhibitor diisopropyl fluorophosphates. GZMB is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp. GZMB seems to be linked to activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution and cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty