Would you like to log in again?
Human Recombinant Il 18Bp (from E. coli)
Supplier: BioVision
AB285830-100UG
10009-798EA 789.07 USD InStock
10009-798
Human Recombinant Il 18Bp (from E. coli)
Proteins and Peptides
≥90% pure human Recombinant IL18BP
IL18BP functions as an inhibitor of the proinflammatory cytokine, IL18. It binds IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production, resulting in reduced T-helper type 1 immune responses. This protein is constitutively expressed and secreted in mononuclear cells. Elevated level of this protein is detected in the intestinal tissues of patients with Crohn's disease. Recombinant human IL18BP protein, fused to His-tag at N-terminus, was expressed in E.coli.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty