Would you like to log in again?
Rat Recombinant IL17A/F (from E. coli)
Supplier: BioVision
AB285915-10UG AB285915-1MG AB285915-50UG
10009-870EA 919.33 USD InStock
10009-870 10009-872 10009-874
Rat Recombinant IL17A/F (from E. coli)
Proteins and Peptides
≥98% pure active rat Recombinant IL17 A/F
The activity determined by the dose-dependent induction of IL-6 production in cultured mouse NIH 3T3 fibroblasts, is typically 20-30 ng/mL.
Interleukin-17AF (IL-17AF) is a member of the IL-17 family of proteins produced by a subset of T cells, called Th17, following stimulation with IL-23. Since IL-17AF is thought to signal through the IL-17R receptor, its biological function is similar to that of IL-17A in that it induces the production of a variety of chemokines, in addition to airway neutrophilia. In regard to these functions, IL-17AF has less activity than the IL-17A homodimer but, greater activity than the IL-17F homodimer. Human and rat IL-17AF both show activity on mouse cells. Recombinant rat IL-17AF is a non-glycosylated, disulfide-linked heterodimer. It is comprised of one monomeric subunit each of IL-17A and IL-17F, having a total of 269 amino acids and a molecular weight of 30.7 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty