Would you like to log in again?
Rat Recombinant Cathepsin (active) (from E. coli)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285935-50UG AB285933-5UG
10009-888EA 5439.2 USD InStock
10009-888 10009-886
Rat Recombinant Cathepsin (active) (from E. coli)
Proteins and Peptides
≥90% pure Active Rat Recombinant Cathepsin K
The specific activity is >500 mU/mg (1U = 1 µmole/min/mg) as measured by Cathepsin K Activity Assay Kit (Catalog #K141-100)
Cathepsin K, a member of the papain cysteine proteinase family is the predominant proteinase responsible for the resorption of the bone matrix. Cathepsin cleaves proteins such as collagen type I, collagen type II and osteonectin, thereby playing a role in bone remodeling and resorption in osteoporosis, osteolytic bone metastasis and rheumatoid arthritis (Bromme and Okamoto, 1995; Drake, F. et al 1996; Bossard et al, 1996).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty