Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant ACT (from Insect cells)
Supplier: BioVision
AB168075-5UG
10009-924EA 574.56 USD InStock
10009-924
Human Recombinant ACT (from Insect cells)
Enzymes
>90% Pure Recombinant Human Beta-Secretase.
BioVision’s recombinant human beta-Secretase/BACE-1 is biologically active as determined by using the β-Secretase Activity Assay Kit (Cat# K360-100).
β-Secretase, also called BACE1 (β-site of APP cleaving enzyme) or memapsin-2 is an aspartic-acid protease important in the pathogenesis of Alzheimer's disease, and in the formation of myelin sheaths in peripheral nerve cells. The transmembrane protein, contains two active site aspartate residues in its extracellular protein domain and may function as a dimer..
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty