Would you like to log in again?
Human Recombinant BRDT (from E. coli)
Supplier: BioVision
AB176961-20UG
10009-320EA 543.92 USD InStock
10009-320
Human Recombinant BRDT (from E. coli)
Proteins and Peptides
>98% Pure Recombinant Human Bromodomain-containing Protein 4 (BrD4) (257-382 aa).
BrdT is similar to the RING3 protein family. It possesses 2 bromodomain motifs and a PEST sequence. The bromodomain is found in proteins that regulate transcription. BrdT drives a meiotic and post-meiotic gene expression program. It also controls the genome-wide post-meiotic genome reorganization that occurs after histone hyperacetylation in elongating spermatids. It may play a role in the transcriptional regulation of spermatogenesis. It also seems to have a structural ATP-independent role in the reorganization of acetylated chromatin. BioVision’s recombinant protein includes Bromo 1 domain (257-382 aa) with N-terminal His-tag.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty