Would you like to log in again?
Human Recombinant BRD4 (from E. coli)
Supplier: BioVision
AB176936-20UG
10009-326EA 550.53 USD InStock
10009-326
Human Recombinant BRD4 (from E. coli)
Proteins and Peptides
>98% Pure Recombinant Human Bromodomain-containing Protein 4 (BrD4) (49-170 aa).
BrD4 protein is homologous to the murine protein MCAP, which associates with chromosomes during mitosis, and to the human RING3 protein. Each of these proteins contains two bromodomains, a conserved sequence motif which may be involved in chromatin targeting. It plays a role in a process governing chromosomal dynamics during mitosis. BioVision’s BrD4 recombinant protein includes Bromo 2 domain (49-170 aa) with an N-terminal His-tag.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty