Would you like to log in again?
Human Recombinant Creb Bp Brd 1081-1197 aa H (from E. coli)
Supplier: BioVision
AB176930-20UG
10009-504EA 474.97 USD InStock
10009-504
Human Recombinant Creb Bp Brd 1081-1197 aa H (from E. coli)
Proteins and Peptides
≥95% Pure Recombinant Human CREB Binding Protein bromodomain (1081-1197 aa)
The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. The cAMP response element-binding protein (CREB) binding protein (CREBBP) bromodomain has been shown to modulate the stability and function of the tumor suppressor protein p53. CREBBP bromodomain recognizes the acetylated lysine residue 382 on p53. This product contains the bromodomain region of CREBBP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty