Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Glucokinase (from E. coli)
Supplier: BioVision
AB123840-10UG
10013-138EA 382.46 USD InStock
10013-138
Human Recombinant Glucokinase (from E. coli)
Enzymes
>80% Pure active Recombinant Pancreatic Glucokinase.
Glucose is phosphorylated to glucose-6-phosphate by glucokinases. This gene is alternatively spliced to generate three different forms of the enzyme; one found in the pancreas and two found in the liver. The main function of this gene is to regulate carbohydrate metabolism. Recombinant human pancreatic Glucokinase (Accession# NP_000153)) has a C-terminal FLAG tag and has 470 amino acid residues. It can be useful for studies including enzyme kinetics, activator screening and kinase selectivity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty