Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Recombinant Turbotev Protease (from E. coli)
Supplier: BioVision
AB285997-1MG
10013-238EA 966.4 USD InStock
10013-238
Recombinant Turbotev Protease (from E. coli)
Enzymes
Pure recombinant active bacterial TurboTEV Protease
BioVision’s TurboTEV Protease is a cysteine protease that recognizes the cleavage site of Glu-Xaa- Xaa-Y- Xaa-Gln-(Gly/Ser) and cleaves between Gln and Gly; the optimal sequence is Glu-Asn-Leu-Tyr-Phe-Gln-Ser. It contains an enhanced form of a catalytic fragment of the N1a protein of Tobacco etch virus (TEV), TurboTEV. TurboTEV Protease is a restriction grade protease that has robust activity at 4°C with high specificity and great stability. It does not require any special buffer for its activity and can be used in a buffer most suitable for the target protein. TurboTEV Protease is a 52 kDa protein with both GST and His tags so it can be removed by either Ni-chelating or glutathione (GSH) resin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty