Would you like to log in again?
Tritirachium album Recombinant Proteinase K
Supplier: BioVision
AB286002-500MG AB286002-100MG AB286002-584
10013-246EA 3123.24 USD InStock
10013-246 10013-244 10013-250
Tritirachium album Recombinant Proteinase K
Enzymes
>90% Pure Recombinant Proteinase K (lyophilized)
A serine protease that displays the ability to digest native proteins, thereby inactivating enzymes such as DNase and RNase without recourse to a denaturation process. It retains its activity in presence of SDS and urea. It is inactivated by diisopropyl fluorophosphates (DFP) and phenyl methane sulfonyl fluoride (PMSF).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty