Would you like to log in again?
Recombinant Chk2 (active) (from Baculovirus (Sf9 Insect cells))
Supplier: BioVision
AB62285-5
10011-976EA 386.92 USD InStock
10011-976
Recombinant Chk2 (active) (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
>90% Pure active Recombinant CHK2.
Chk2 is the mammalian homolog of the Saccharomyces cerevisiae Rad53 and Schizosaccharomyces pombe Cds1 protein kinases required for the DNA damage and replication checkpoints. Chk2 is rapidly phosphorylated and activated in response to replication blocks and DNA damage; the response to DNA damage occurs in an ataxia telangiectasia mutated (ATM)-dependent manner (1). In vitro, Chk2 phosphorylates p53 on Ser-20 and dissociated preformed complexes of p53 with Mdm2, a protein that targets p53 for degradation (2). In vivo, ectopic expression of wild-type Chk2 leads to increased p53 stabilization after DNA damage, whereas expression of a dominant-negative Chk2 mutant abrogated both phosphorylation of p53 on Ser-20 and p53 stabilization. Thus, in response to DNA damage, Chk2 stabilizes the p53 tumor suppressor protein leading to cell cycle arrest in G1. Chk2 is directly phosphorylated by ATM in response to ionizing radiation (3). The phosphorylation occurs in the Ser-Gln/Thr-Gln (SQ/TQ) cluster domain (SCD) on Chk2, which contains seven SQ/TQ motifs, and Thr68 is the major in vitro phosphorylation site by ATM.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty