Would you like to log in again?
Human Protein G Sepharose
Supplier: BioVision
AB193259-25ML AB193259-100ML AB193259-1ML AB193259-5ML
10008-882EA 1427.04 USD InStock
10008-882 10008-880 10008-878 10008-876
Human Protein G Sepharose
Proteins and Peptides
Protein G Sepharose for High Specific Binding of IgG (> 20 mg human or rabbit IgG/ml Protein G-Sepharose). High Flow Rate, Minimal leaching of the ligand. Reusable for up to 10 times without significant loss of binding capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty