Would you like to log in again?
Hi-Bind™ Human G-Agarose
Supplier: BioVision
Hi-Bind™
AB193258-25ML AB193258-100ML AB193258-5ML AB193258-1ML
10008-894EA 1347.96 USD InStock
10008-894 10008-892 10008-890 10008-888
Hi-Bind™ Human G-Agarose
Proteins and Peptides
Protein G Agarose for High Specific Binding of IgG (≥ 30 mg human or rabbit IgG/ml Protein G-Agarose). High Flow Rate, Minimal Leaching of the ligand. Reusable for up to 10 times without significant loss of binding capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty