Would you like to log in again?
Protein G Sepharose Columns, BioVision
Supplier: BioVision
AB193260-5ML AB193260-1ML
10008-900EA 1667.53 USD InStock
10008-900 10008-898
Protein G Sepharose Columns, BioVision
Chromatography Columns
Ready-to-use pre-packed columns with reusable Protein G conjugated Sepharose beads for antibody purification from multiple sources.
  • Minimal leaching of the ligand
Reusable for up to 10 times without significant loss of binding capacity.
Applications: Purification of monoclonal and polyclonal antibodies from culture media, serum, ascites fluid or hybridoma supernatants. Isolation of antibody/antigen complexes in immunoprecipitation experiments, since only the Fc region is involved in antibody binding and the Fab region is available for binding antigen.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty