Would you like to log in again?
Protein A/G Sepharose Columns, BioVision
Supplier: BioVision
AB193263-1ML AB193263-5ML
10008-922EA 1904.65 USD InStock
10008-922 10008-920
Protein A/G Sepharose Columns, BioVision
Chromatography Columns
Ready-to-use pre-packed columns with reusable Protein A/G conjugated Sepharose beads for antibody purification from multiple sources.
  • High flow rate
  • Low falling off of rProteinA/G
Applications: Purification of monoclonal and polyclonal antibodies.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty