Would you like to log in again?
Gsk-3A-Gst Fusion (from E. coli), BioVision
Supplier: BioVision
AB285748-100UG
10008-814EA 306.43 USD InStock
10008-814
Gsk-3A-Gst Fusion (from E. coli), BioVision
Proteins and Peptides
Pure active Human Recombinant GSK-3a-GST Fusion Protein
GSK-3 is one of the Akt substrates. It can be phosphorylated by Akt at Ser21 of GSK-3α or Ser9 of GSK-3β. The phosphorylation of GSK negatively regulates its kinase activity. The recombinant GSK-3α fusion protein serves as a useful substrate for assaying Akt kinase activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty