Would you like to log in again?
Human Native Kallikrein (from Plasma)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285481-50UG
10005-614EA 329.4 USD InStock
10005-614
Human Native Kallikrein (from Plasma)
Proteins and Peptides
>95% Pure Kallikrein, Human Plasma.
A serine protease that releases bradykinin from kininogen. Present in plasma as an inactive precursor prokallikrein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty