Would you like to log in again?
Heparin Sepharose Columns, BioVision
Supplier: BioVision
AB286844-5ML AB286844-1ML
10008-970EA 410.76 USD InStock
10008-970 10008-972
Heparin Sepharose Columns, BioVision
Chromatography Columns
Ready-to-use pre-packed columns with high binding capacity beads for affinity purification of heparin binding proteins.
  • Excellent binding capacity
  • High flow rate
  • No significant loss of the heparin ligand
  • >2.5 mg heparin per ml Sepharose beads
Heparin-beads have been widely used in affinity purification of various heparin-binding proteins or ligand, such as antithrombin III, lipoprotein, DNA binding proteins (transcription factors, virus coat proteins and more.) and other ligands. BioVison’s Heparin-Sepharose is designed for the purpose of purification of these kinds of proteins and ligands. It can also be used as a high capacity cation exchange medium. Specific proteins can be separated by using different concentrations of salt or a salt gradient.
Applications: Purification of heparin-binding proteins, enzymes or other ligands.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty