Would you like to log in again?
Human Jacalin-Sepharose
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB193266-1ML AB193266-5ML AB193266-25ML
10008-990EA 566.94 USD InStock
10008-990 10008-988 10008-986
Human Jacalin-Sepharose
Proteins and Peptides
Jacalin-Sepharose for High Specific Binding of IgA (≥ 3 mg human IgA/ml Jacalin-Sepharose). High Flow Rate, Minimal leaching of the Ligand.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty