Would you like to log in again?
Hi-Bind™ Ni Qr Agarose Beads
Supplier: BioVision
Hi-Bind™
AB286860-100ML AB286860-10ML AB286860-1ML AB286860-500ML
10008-992EA 137.9 USD InStock
10008-992 10008-998 10008-996 10008-994
Hi-Bind™ Ni Qr Agarose Beads
Proteins and Peptides
Hi-Bind™ Ni QR Agarose Beads: for specific purification of native and recombinant proteins and peptides that have an affinity for metal ions (up to 60 mg target protein/ml of settled resin), high flow rate & convenient pack sizes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty