Would you like to log in again?
Streptavidin-Sepharose Beads, Biovision
Supplier: BioVision
AB286845-10ML AB286845-5ML AB286845-2ML
10009-002EA 1078.55 USD InStock
10009-002 10009-004 10009-006
Streptavidin-Sepharose Beads, Biovision
Protein Purification Protein Purification Resins
Streptavidin Sepharose beads can be used for the purification of biotinylated biomolecules for subsequent ELISA, immunoassays and other downstream analyses. High binding capacity and flexible sizing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty