Would you like to log in again?
Human Core Histones (from HeLa S3 (cervical adenocarcinoma))
Supplier: BioVision
AB176964-50UG
10011-060EA 712.41 USD InStock
10011-060
Human Core Histones (from HeLa S3 (cervical adenocarcinoma))
Proteins and Peptides
≥95% Pure Human Core Histones
A nucleosome is the basic repeating unit of chromatin in which 146 base pairs of DNA wrap twice around a histone octamer consisting of two copies of each of the core histones, H2A, H2B, H3, and H4. The combination of two H2A/H2B dimers and one H3/H4 tetramer create the nucleosome core.2 Histone H3 undergoes many modifications which include acetylation, methylation, and phosphorylation that are important for regulation of gene transcription.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty