Would you like to log in again?
Virgo® EBV-VCA ELISA Test System, Hemagen
Supplier: Hemagen Diagnostics
Virgo®
6619 6618
10818-918KT 765.2 USD InStock
10818-918 10818-920
Virgo® EBV-VCA ELISA Test System, Hemagen
Assays ELISAs
For the detection and titration of EBV-viral capsid antigen (VCA) IgG and IgM antibodies in human sera.
  • Antibody Kit
  • FDA Compliant
  • For detection and titration
Certifications: FDA Compliant
Ordering information: Accessories included.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty