Would you like to log in again?
OxiSelect™ Superoxide Dismutase Activity Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-340
102964-966EA 726.15 USD InStock
102964-966
OxiSelect™ Superoxide Dismutase Activity Assay Kit, Cell Biolabs
Assays Cellular Assays
OxiSelect™ Superoxide Dismutase Activity Assay uses a xanthine/xanthine oxidase (XOD) system to generate superoxide anions and a chromagen to produce a water-soluble formazan dye upon reduction by superoxide anions
The superoxide dismutase activity is determined as the inhibition or reduction of chromagen. This kit is suitable for use with plasma, serum, cells or tissues. A superoxide dismutase standard included for absolute quantitation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty