Would you like to log in again?
OxiSelect™ Hydrogen Peroxide/Peroxidase Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-344 STA-844
102965-320EA 590.77 USD InStock
102965-320 102965-792
OxiSelect™ Hydrogen Peroxide/Peroxidase Assay Kits, Cell Biolabs
Assays Enzyme Assays
The OxiSelect™ Hydrogen Peroxide/Peroxidase Assay Kits are sensitive quantitative assays for detecting hydrogen peroxide or peroxidase.  Kits are available with either colorimetric or fluorometric detection in a plate reader. In the presence of peroxidase, a probe reacts with H₂O₂ in a 1:1 stoichiometry to produce a colored product or fluorescent product. Readings are proportional to the H₂O₂ or peroxidase levels within the samples, and the H₂O₂ or peroxidase content in unknown samples is determined by comparison with the respective standard curve.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty