Would you like to log in again?
Nuclear/Cytosolic Fractionation Kits, Cell Biolabs
Supplier: Cell Biolabs
AKR-172 AKR-171
102963-636EA 530 USD InStock
102963-636 102963-634
Nuclear/Cytosolic Fractionation Kits, Cell Biolabs
Assays Cellular Assays
Cell Biolabs’ Nuclear/Cytosolic Fractionation Kit provides a simple and fast tool to isolate nuclear extract from the cytoplasmic fraction of mammalian cells.  The procedure has been optimized to provide extraction, with high protein recovery and low cross-contamination, in less than 2 hours.  The extracted protein fractions are functional and suitable for downstream assays such as DNA footprinting, RNA splicing, gel shift assays (EMSA), reporter assays, enzyme activity assays, and Western blotting
Each prep can accommodate up to 5 million cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty