Would you like to log in again?
Uric Acid/Uricase Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-375
102965-042EA 726.15 USD InStock
102965-042
Uric Acid/Uricase Assay Kit, Cell Biolabs
Assays Immunological Assays
Cell Biolabs’ Uric Acid/Uricase Assay Kit is a simple HTS-compatible assay for measuring uric acid concentrations in biological samples such as serum, plasma, and urine without any need for pretreatment.  The kit has a detection sensitivity limit of 0.5 µM of uric acid or 1 mU/mL of uricase
 Each kit provides sufficient reagents to perform up to 400 assays, including standard curve and unknown samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty