Would you like to log in again?
CytoSelect™ 24-Well Cell Co-Culture System, Cell Biolabs
Supplier: Cell Biolabs
CBA-160 CBA-160-5
102963-826EA 2067.69 USD InStock
102963-826 102963-824
CytoSelect™ 24-Well Cell Co-Culture System, Cell Biolabs
Assays Cellular Assays
CytoSelect™ 24-Well Cell Co-Culture System provides a unique platform to monitor direct contact between two cell types in a single well.  The kit uses a proprietary molded insert that creates a gap in the center of the well
The first cell type is cultured until a monolayer forms around the insert, creating a defined 8 mm cell-free zone.  Once the insert is removed, a second cell type may be seeded into the exposed zone.  The kit contains proprietary treated inserts and sufficient reagents for the evaluation of 24 samples.  The inserts are compatible with most adherent cell types and experimental conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty