Would you like to log in again?
CytoSelect™ 96-Well Endothelium & Epithelium Adhesion Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
CBA-210 CBA-215 CBA-211
102966-502EA 475 USD InStock
102966-502 102966-498 102966-496
CytoSelect™ 96-Well Endothelium & Epithelium Adhesion Assay Kits, Cell Biolabs
Assays Cellular Assays
CytoSelect™ 96-Well Endothelium & Epithelium Adhesion Assays provide a convenient plate-based method for the quantitation of interactions between cells and the endothelium or epithelium
No manual cell counting is required. Quantitation is achieved by a fluorescent dye included in each assay kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty