Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG2 beta Goat Polyclonal Antibody (IRDye® 680LT)

Supplier: LI-COR®
926-68052
102673-406EA 380.31 USD InStock
102673-406
Anti-IgG2 beta Goat Polyclonal Antibody (IRDye® 680LT)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgG2b covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy chain of mouse IgG2b. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with mouse IgM, IgG1, IgG2a, IgG3, IgA, pooled human sera, and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: Western blot, proein array, immunohistochemistry, microscopy, 2D gel detection, and tissue section imaging. NOT RECOMMENDED for: small animal imaging, In-Cell Wesern assays, or On-Cell Western assays.

Western blot: 1:20000

Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope:
Host: Goat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty