Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)

Supplier: LI-COR®
925-32214
102970-996EA 146.46 USD InStock
102970-996
Anti-IgG Donkey Polyclonal Antibody (IRDye® 800CW)
Antibodies
The antibody was isolated from antisera by affinity chromatography using antigens coupled to agarose beads. Based on immunoelectrophoresis, the antibody reacts with the heavy chains on goat IgG and with the light chains common to most goat immunoglobulins. The antibody also reacts with sheep IgG. No reactivity was detected against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reactivity with chicken, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and rat serum proteins, but the antibody may cross-react with immunoglobulins from other species. The conjugate was specifically tested and qualified for Western blot applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty