Would you like to log in again?
Urea Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-382
102965-052EA 726.15 USD InStock
102965-052
Urea Assay Kit, Cell Biolabs
Assays Immunological Assays
Cell Biolabs’ Urea Assay Kit is based on the Berthelot reaction.  Urea is first degraded into ammonia and carbon dioxide, which further reacts with an alkaline developer to produce a blue-green colored product that can be measured with a standard spectrophotometric plate reader at an optical density between 580-630 nm.  Each kit provides sufficient reagents to perform up to 192 assays, including blanks, urea standards and unknown samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty