Would you like to log in again?
Human Albumin ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-383
102965-054EA 695.38 USD InStock
102965-054
Human Albumin ELISA Kit, Cell Biolabs
Assays ELISAs
Cell Biolabs’ Human Albumin ELISA Kit is an enzyme immunoassay developed for the detection and quantitation of human albumin in plasma, serum, urine or other biological fluid samples. The kit has a detection sensitivity limit of 5 ng/mL of human albumin.  Each kit provides sufficient reagents to perform up to 96 assays including standard curve and unknown samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty