Would you like to log in again?
Total Cholesterol Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-384 STA-390
102965-068EA 566.15 USD InStock
102965-068 102965-056
Total Cholesterol Assay Kits, Cell Biolabs
Assays Immunological Assays
Cell Biolabs’ Total Cholesterol Assay Kits measure the total cholesterol within serum, plasma, cell lysate, or tissue samples
The assays will detect total cholesterol (cholesteryl esters plus free cholesterol) in the presence of cholesterol esterase or only free cholesterol in the absence of the esterase enzyme. Kits are available with either colorimetric or fluorometric detection in a 96-well plate reader.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty