Would you like to log in again?
CytoSelect™ BrdU Cell Proliferation ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
CBA-251
102963-848EA 584.62 USD InStock
102963-848
CytoSelect™ BrdU Cell Proliferation ELISA Kit, Cell Biolabs
Assays ELISAs
The CytoSelect™ BrdU Cell Proliferation ELISA Kit detects BrdU incorporated into cellular DNA during cell proliferation using an anti-BrdU antibody
When cells are incubated in media containing BrdU, the pyrimidine analog is incorporated in place of thymidine into the newly synthesized DNA of proliferating cells. Once the labeling media is removed, the cells are fixed and the DNA is denatured in one step with a fix/denature solution (denaturation of the DNA is necessary to improve the accessibility of the incorporated BrdU for detection). Then an anti-BrdU mouse monoclonal antibody is added followed by an HRP conjugated secondary antibody to detect the incorporated BrdU. The magnitude of the absorbance for the developed color is proportional to the quantity of BrdU incorporated into cells and can be directly correlated to cell proliferation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty