Would you like to log in again?
Glutamate Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-674
102966-248EA 572.31 USD InStock
102966-248
Glutamate Assay Kit, Cell Biolabs
Assays Enzyme Assays
Cell Biolabs' Glutamate Assay Kit is a quantitative, fluorometric assay that uses glutamate specific enzymes to generate hydrogen peroxide
A fluorometric probe is oxidized by hydrogen peroxide to generate fluorescent Resorufin, which correlates to the level of glutamate in the sample. Glutamate levels in an unknown sample are calculated based on a glutamate standard curve.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty