Would you like to log in again?
CellTag™ 700 Stain In-Cell Western™ Products, LI-COR®
Supplier: LI-COR®
926-41090
102971-094EA 220.31 USD InStock
102971-094
CellTag™ 700 Stain In-Cell Western™ Products, LI-COR®
Stains In vivo Imaging Dyes
CellTag 700 Stain is a near-infrared fluorescent, non-specific cell stain that provides accurate normalization to cell number for In-Cell Western™ Assay applications.
  • Accumulates in both the nucleus and cytoplasm of permeabilized cells
  • Provides linear fluorescent signal across a wide range of cell types and cell numbers
  • Detected in the 700 nm channel of Odyssey® CLx, Classic, and Sa Imaging Systems
  • Applied to the cells during incubation
  • Enables accurate measurement of target protein levels
The stain accumulates in both the nucleus and cytoplasm of permeabilized cells, and provides linear fluorescent signal across a wide range of cell types and cell numbers.
CellTag 700 Stain is detected in the 700 nm channel of Odyssey® CLx, Classic, and Sa Imaging Systems. CellTag 700 Stain is applied to the cells during incubation with an IRDye® 800CW secondary antibody, and enables accurate measurement of target protein levels with much higher throughput than Western blotting. CellTag 700 Stain ICW Kits provide detection reagents for cell-based In-Cell Western Assays.
Ordering information: Each kit includes blocking buffer, IRDye® 800CW secondary antibody for detection of a specific protein target in the 800 nm channel, and CellTag 700 Stain to normalize well-to-well variations in cell number. This cost-effective normalization method makes quantification of the target protein more precise.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty