Would you like to log in again?
OxiSelect™ Intracellular Nitric Oxide (NO) Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-800-5 STA-800
102965-258EA 2307.69 USD InStock
102965-258 102965-256
OxiSelect™ Intracellular Nitric Oxide (NO) Assay Kit, Cell Biolabs
Assays Cellular Assays
OxiSelect™ Intracellular Nitric Oxide Assay Kit is a cell-based assay for rapid detection of intracellular NO or NOS activity in cultured cells.  In brief, the cell-permeant NO probe passively diffuses into cells and is deacetylated by cellular esterases to a non-fluorescent intermediate.  When intracellular nitric oxide encounters this intermediate, it rapidly oxidizes to a highly fluorescent, triazolo-fluorescein analog.  The fluorescence intensity is proportional to the NO levels within the cell cytosol (detection limit of about 3 nM). 
The kit’s provided Nitric Oxide Fluorometric Probe is NO specific, has excellent photostablity and pH stability, and is detected with standard fluorescein filters.  The probe is suitable for flow cytometry, fluorescent microscopy, and fluorescent microplate detection.  Each kit provides sufficient reagents to perform up to 96 assays, including blanks, standards, and unknown samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty