Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Hartmann Ear Speculum, OR Grade, Sklar
Supplier: Sklar
67-6004
10198-408EA 52.07 USD InStock
10198-408
Hartmann Ear Speculum, OR Grade, Sklar
Specula
The Sklar Hartman Ear Speculum is a cone/funnel-shaped attachment for an otoscope that allows examination of the ear canal and eardrum
The speculum is inserted into the ear canal and the otoscope handle provides a light source and low-power magnification so the practitioner can view the eardrum. Disease and infections of the middle or inner ear can largely be detected by this examination. This device is a round attachment with a 4mm diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty