Would you like to log in again?
AAV Expression Vectors, Cell Biolabs
Supplier: Cell Biolabs
VPK-411 VPK-417 VPK-419 VPK-430 VPK-416 VPK-418 VPK-410
102965-560EA 515 USD InStock
102965-560 102965-528 102965-514 102965-468 102965-500 102965-542 102965-462
AAV Expression Vectors, Cell Biolabs
Vectors, Plasmids and Libraries
Cell Biolabs' AAV Expression Vectors are designed to work with any AAV packaging system including the AAV Helper Free Packaging Systems
The shRNA expression vectors contain a U6 promoter. All other expression vectors contain a CMV promoter which is optimal for general gene expression, with the exception of our promoterless vector which is useful if you wish to clone in a gene-specific promoter along with your gene of interest.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty