Would you like to log in again?
Littauer Ear Forceps, OR Grade, Sklar
Supplier: Sklar
67-1052
10198-276EA 549.03 USD InStock
10198-276
Littauer Ear Forceps, OR Grade, Sklar
Forceps
Sklar's Littauer Forceps are a non-ratcheted, finger ring instrument commonly used in ear and nasal procedures
This product has short shanks that angle at the screw lock to a 3" long shaft. The jaws are serrated and useful for grasping at tissue or foreign objects that may not be easily seen or reached in the ear or nasal canal.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty