Would you like to log in again?
OxiSelect™ Total Glutathione (GSSG/GSH) Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-312
102964-884EA 578.46 USD InStock
102964-884
OxiSelect™ Total Glutathione (GSSG/GSH) Assay Kit, Cell Biolabs
Assays Enzyme Assays
The OxiSelect™ Total Glutathione Assay Kit is a quantitative assay for measuring the total glutathione content within a sample (GSH and GSSG).  Glutathione Reductase reduces oxidized glutathione (GSSG) to reduced glutathione (GSH) in the presence of NADPH.  Subsequently, the  chromogen reacts with the thiol group of GSH to produce a colored compound that absorbs at 405 nm.  The total glutathione content in unknown samples is determined by comparison with the predetermined glutathione standard curve.  The rate of chromophore production is proportional to the concentration of glutathione within the sample.  Metaphosphoric acid is provided to remove interfering proteins or enzymes from samples
The kit detects glutatione levels as low as 4 nM.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty