Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREX® 75° Angle Tube Adapter, Corning

Supplier: Corning
PYREX®
8980-24
62945-010CS 814.17 USD InStock
62945-010
PYREX® 75° Angle Tube Adapter, Corning
Adapters Adapters, Joints with Tube Connectors
Angle tube adapters with two-way standard taper joints and a standard taper outer joint made of PYREX® borosilicate glass.

  • The sidearm is at an angle approximately 75° from the lower joint
  • The lower end and sidearm are equipped with inner standard taper 24/40 joints
  • The 10/30 outer joint accommodates a thermometer

Length: ~160 mm

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty