Would you like to log in again?
PYREX® Dispensing Burets, Colored Scale, with Straight Bore PTFE Stopcock, Corning
Supplier: Corning
PYREX®
2094-1L 2094-500 2094-250
17475-229CS 329.06 USD InStock
17475-229 17475-207 17475-241
PYREX® Dispensing Burets, Colored Scale, with Straight Bore PTFE Stopcock, Corning
Burets
Burets are mainly designed to dispense large volumes of liquids rapidly and accurately. Suitable for use in school and institutional laboratories not requiring the accuracy of Class A models.
  • Made from close tolerance, accurate bore tubing with uniform walls
  • Tips carefully drawn and ground to reduce chipping
  • With permanent coloured scale
Provided with a PTFE stopcock plug to reduce freezing and eliminate lubricant contamination. The smooth micro-finish of the stopcock barrel assures a precision fit and optimum leak-resistant performance.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty