Would you like to log in again?
PYREX® Filter Tube Funnels, Corning
Supplier: Corning
PYREX®
9480-32 9480-36 9480-27 9480-42 9480-24
28680-069CS 36.21 USD InStock
28680-069 28680-047 28680-025 28680-105 28680-073
PYREX® Filter Tube Funnels, Corning
Funnels Filtration Funnels Filter Funnels
Made from PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Filter tubes have a constricted lower end. They can be used on the bottom end of 30 mm O.D. PYREX® Gooch low form fritted disc crucibles to collect the filtrate. They can also be used directly with porcelain plates.
The constricted lower end is approximately 75 mm long by 8 mm O.D. on all sides.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty