Would you like to log in again?
TC-100 Insect Medium, HiMedia Laboratories
Supplier: HiMedia Laboratories
IM007-20L IM007-5L IM007-10L IML007-5X500ML IM007-50L IM007-1L IM007-1L IM007-5L IM007-50L IM007-10L IML007-2X500ML IM007-20L
95037-460PK 149.31 USD InStock
95037-460 95038-654 CA95039-076 95038-652 CA95038-652 95039-076 CA95039-074 CA95038-654 95037-458 CA95039-072 95039-072 95039-074
TC-100 Insect Medium, HiMedia Laboratories
Cell Culture Media
TC-100 is a modification done by Dr Gardiner and Dr Stockdale of the original formulation of Grace’s insect medium.
  • Modification to optimise production of Autographia californica NPV virions by cells from the fall armyworm
TC-100 is a modification done by Dr Gardiner and Dr Stockdale of the original formulation of Grace’s insect medium. Sucrose, fructose and several krebs’s cycle intermediates were omitted from the original formula. The modification was done to optimise the production of Autographia californica NPV virions by cells from the fall armyworm, Spodoptera frugiperda.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty