Would you like to log in again?
Basal Medium Eagle (BME) Mammalian Cell Culture Medium, HiMedia
Supplier: HiMedia Laboratories
AT002-1L AT061-5L AT061-5L AT061-10L AT001-20L AT061-1L AT001-1L AT061-1L AT001-5L AT001-5L AT061-10L AT002-1L AT002-20L AT001-1L AT002-5L AT002-5L AT001-20L AT002-20L
95016-440EA 42.09 USD InStock
CA95016-440 95016-440 95016-432 95016-442 CA95016-444 95016-434 CA95016-434 95016-444 CA95016-442 95016-436 CA95016-432 95016-446 95016-438 CA95016-448 CA95016-438 95016-448 CA95016-446 CA95016-436
Basal Medium Eagle (BME) Mammalian Cell Culture Medium, HiMedia
Cell Culture Media
Basal Medium developed by Harry Eagle is a combination of essential nutrients in appropriate concentrations for monolayer cultivation of a wide variety of normal and transformed cells.
  • Initially developed to determine the nutritional requirements of Hela cells and mouse fibroblast L cells
  • Name applies to only the formulation developed for HeLa cells
  • Supports growth of variety of diploid or primary mammalian cell cultures
  • Modifications result in other media
The medium was initially developed as a result of studies to determine the nutritional requirements of Hela cells and mouse fibroblast L cells in culture. Although there are many versions of basal medium described by Eagle, the name ‘Basal Medium, Eagle’ applies to only the formulation developed for HeLa cells. Basal medium eagle when properly supplemented supports growth of variety of diploid or primary mammalian cell cultures. Modifications to the original BME have resulted in other media, including MEM and DMEM.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty