Would you like to log in again?
Leibovitz's L-15 Medium Mammalian Cell Culture Medium, HiMedia Laboratories
Supplier: HiMedia Laboratories
AT011-1L AT207-10X1L AT011-20L AT207-20L AT011-5L AT207-5L AT207-1L AT011-20L AT011-1L AT207-5L AT207-10X1L AT207-20L AT011-5L AT207-1L
89222-116EA 30.26 USD InStock
89222-116 CA89222-114 89222-114 CA89222-118 89222-118 CA89222-116 89222-120 CA89222-120 95016-512 CA95016-514 95016-514 CA95016-512 95016-516 CA95016-516
Leibovitz's L-15 Medium Mammalian Cell Culture Medium, HiMedia Laboratories
Cell Culture Media
Leibovitz Medium was specifically designed to grow cells in a CO2 free atmosphere.
  • Does not require supplementation with sodium bicarbonate
  • Can be used under conditions of free gaseous exchange with the atmosphere
  • Can be used to grow human tumor cells and embryonic cells
  • Frequently used in diagnostic virology
The standard sodium bicarbonate/CO2 buffering system is replaced by a combination of free base amino acids, phosphate buffers, and higher levels of galactose and sodium pyruvate. As a result, the medium does not require supplementation with sodium bicarbonate and can be used under conditions of free gaseous exchange with the atmosphere. The medium can be used to grow human tumor cells and embryonic cells and also established cell lines like HeLa and Hep-2. The medium is frequently used in diagnostic virology where tissue cell lines or strains need to be grown in closed systems. Leibovitz medium obviates the need for frequent medium change.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty